Skip to Content
MilliporeSigma
All Photos(2)

Documents

N28353

Sigma-Aldrich

1,3-Dimethyl-2-nitrobenzene

99%

Synonym(s):

2-Nitro-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
2046066
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.521 (lit.)

bp

225 °C/744 mmHg (lit.)

mp

14-16 °C (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)c1[N+]([O-])=O

InChI

1S/C8H9NO2/c1-6-4-3-5-7(2)8(6)9(10)11/h3-5H,1-2H3

InChI key

HDFQKJQEWGVKCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung Seok Lee et al.
Macromolecular bioscience, 15(9), 1314-1322 (2015-06-04)
We designed poly(β-amino esters) (PBAEs) bearing both UV light- and pH-sensitive groups and used PBAEs to prepare nanoparticles (NPs) that can be utilized for on-demand burst release of guest molecules in response to multiple triggers. Due to the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service