Skip to Content
MilliporeSigma
All Photos(1)

Documents

171018

Sigma-Aldrich

2,4-Dimethyl-1-nitrobenzene

98%

Synonym(s):

1,3-Dimethyl-4-nitrobenzene, 4-Nitro-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
1865656
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%
98%

refractive index

n20/D 1.549 (lit.)

bp

244 °C (lit.)

mp

7-9 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(c(C)c1)[N+]([O-])=O

InChI

1S/C8H9NO2/c1-6-3-4-8(9(10)11)7(2)5-6/h3-5H,1-2H3

InChI key

BBUPBICWUURTNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dimethyl-1-nitrobenzene was used in the synthesis of 1-alkynylphosphines and its oxides.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

248.0 - 249.8 °F - Pensky-Martens closed cup

Flash Point(C)

120 - 121 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-catalyzed synthesis of 1-alkynylphosphine oxides from 1-alkynes and tetraphenylbiphosphine.
Arisawa M, et al.
Tetrahedron Letters, 47(29), 5211-5213 (2006)
Tomohiro Ito et al.
Environmental science and pollution research international, 26(22), 22747-22755 (2019-06-07)
Secondary organic aerosol (SOA) is a component of airborne particulate matter in urban areas. However, their toxicities remain incompletely understood. In this study, we investigated the oxidative and inflammatory potency of SOA derived from three different volatile organic compounds (α-pinene
Tien D Ho et al.
Journal of chromatography. A, 1361, 217-228 (2014-08-26)
Ionic liquids (ILs) were used as a new class of diluents for the analysis of two classes of genotoxic impurities (GTIs), namely, alkyl/aryl halides and nitro-aromatics, in small molecule drug substances by headspace gas chromatography (HS-GC) coupled with electron capture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service