Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

194921

Sigma-Aldrich

N-(2,6-Dimethylphenyl)chloroacetamide

99%

Synonym(s):

1-Chloroacetylamino-2,6-dimethylbenzene, 2-Chloro-2′,6′-acetoxylidide, 2-Chloro-2′,6′-dimethylacetanilide, 2-Chloro-N-(2,6-dimethylphenyl)acetamide, N-(2,6-Dimethylphenyl)-2-chloroacetamide, N-(2,6-Xylyl)chloroacetamide, N-Chloroacetyl-2,6-dimethylaniline, Chloroacetamido-2,6-xylidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONHC6H3(CH3)2
CAS Number:
Molecular Weight:
197.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

mp

150-151 °C (lit.)

functional group

amide
chloro

SMILES string

Cc1cccc(C)c1NC(=O)CCl

InChI

1S/C10H12ClNO/c1-7-4-3-5-8(2)10(7)12-9(13)6-11/h3-5H,6H2,1-2H3,(H,12,13)

InChI key

FPQQSNUTBWFFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

N-(2,6-Dimethylphenyl)chloroacetamide (2-chloro-2′,6′-acetoxylidide) is nonsteroidal antinflammatory drug[1].

Application

N-(2,6-Dimethylphenyl)chloroacetamide (2-chloro-2′,6′-acetoxylidide) was used in simultaneous determination of the enantiomers of tocainide in blood plasma by gas-liquid chromatography with electron-capture detection[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Sedman et al.
Journal of chromatography, 306, 155-164 (1984-03-09)
Tocainide is a new experimental antiarrhythmic agent used clinically as the racemic mixture of two enantiomers. Since the optical isomers may differ in their efficacy and toxicity, we have initiated studies on the stereoselective disposition of tocainide. For this purpose
Application of molecular topology to the search of novel NSAIDs: experimental validation of activity.
Galvez-Llompart M, et al.
Letters in Drug Design & Discovery, 7(6), 438-445 (2010)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service