Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N12821

Sigma-Aldrich

4-Nitrobenzyl alcohol

99%

Synonym(s):

(4-Nitrophenyl)methanol, 1-(Hydroxymethyl)-4-nitrobenzene, 4-(Hydroxymethyl)nitrobenzene, 4-Nitrobenzenemethanol, p-(Hydroxymethyl)nitrobenzene, p-Nitrobenzyl alcohol, p-Nitrophenylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
1424026
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

bp

185 °C/12 mmHg (lit.)

mp

92-94 °C (lit.)

SMILES string

OCc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H7NO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4,9H,5H2

InChI key

JKTYGPATCNUWKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-Kyung Kim et al.
Nucleic acids research, 45(5), 2865-2874 (2016-11-02)
Programmed -1 ribosomal frameshifting (-1PRF) is tightly regulated by messenger RNA (mRNA) sequences and structures in expressing two or more proteins with precise ratios from a single mRNA. Using single-molecule fluorescence resonance energy transfer (smFRET) between (Cy5)EF-G and (Cy3)tRNALys, we
Janghyun Yoo et al.
Nature communications, 11(1), 3336-3336 (2020-07-06)
We describe theory, experiments, and analyses of three-color Förster resonance energy transfer (FRET) spectroscopy for probing sub-millisecond conformational dynamics of protein folding and binding of disordered proteins. We devise a scheme that uses single continuous-wave laser excitation of the donor
Roger B Altman et al.
Nature methods, 9(1), 68-71 (2011-11-15)
Fluorescence applications requiring high photostability often depend on the use of solution additives to enhance fluorophore performance. Here we demonstrate that the direct or proximal conjugation of cyclooctatetraene (COT), 4-nitrobenzyl alcohol (NBA) or Trolox to the cyanine fluorophore Cy5 dramatically
K D James et al.
Journal of bacteriology, 182(11), 3136-3141 (2000-05-16)
Pseudomonas sp. strain TW3 is able to metabolize 4-nitrotoluene to 4-nitrobenzoate and toluene to benzoate aerobically via a route analogous to the upper pathway of the TOL plasmids. We report the cloning and characterization of a benzyl alcohol dehydrogenase gene
Sijia Peng et al.
The journal of physical chemistry. B, 122(18), 4844-4850 (2018-04-19)
Fluorescence correlation spectroscopy is a powerful single-molecule tool that is able to capture kinetic processes occurring at the nanosecond time scale. However, the upper limit of its time window is restricted by the dwell time of the molecule of interest

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service