191396
3-Aminobenzyl alcohol
97%
Synonym(s):
3-(Hydroxymethyl)aniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
92-95 °C (lit.)
SMILES string
Nc1cccc(CO)c1
InChI
1S/C7H9NO/c8-7-3-1-2-6(4-7)5-9/h1-4,9H,5,8H2
InChI key
OJZQOQNSUZLSMV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis and fungicidal activity of N-[4-(4-fluoro) phenyl-2-piperidin-4-yl-thiazol-5-yl] pyrimidin-2-yl-N-phenylamines on Phytophthora capsici.
Journal of the Korean Society for Applied Biological Chemistry, 53(2), 206-214 (2010)
Biotechnology progress, 22(1), 326-330 (2006-02-04)
A new approach to galacto-oligosaccharides and galacto-conjugates synthesis performed by the beta-galactosidase from Kluyveromyces lactis is reported. The enzymatic galactosylation of eight kinds of adsorbed aromatic primary alcohols, in particular the two drugs guaifenesin and chlorphenesin, gave the corresponding beta-D-galacto-pyranosides
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(11), 2511-2517 (2003-09-10)
The laser Raman and FTIR spectra of 3-aminobenzyl alcohol have been recorded. The observed frequencies were assigned to various modes of vibrations on the basis of normal coordinate analysis, assuming C(s) point group symmetry. The potential energy distribution associated with
Synthesis and characterization of new 2-(alkylamino) acetamides.
ARKIVOC (Gainesville, FL, United States), 11, 37-47 (2003)
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 175, 76-91 (2016-12-27)
In this paper three different "push-pull" 4-aminoazobenzene dyes have been synthesized in order to characterize their photochromic behavior in different solvents. The molecular geometry was optimized by DFT/B3LYP functional combined with the standard 6-31+G(d,p) basis set for trans (E) and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service