Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

127809

Sigma-Aldrich

4-Methylbenzyl alcohol

98%

Synonym(s):

(4-Methylphenyl)methanol, (4-Tolyl)methanol, 4-Methylbenzenemethanol, p-Methylbenzyl alcohol, p-Tolualcohol, p-Tolylcarbinol, p-Tolylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2OH
CAS Number:
Molecular Weight:
122.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

217 °C (lit.)

mp

59-61 °C (lit.)

functional group

hydroxyl

SMILES string

Cc1ccc(CO)cc1

InChI

1S/C8H10O/c1-7-2-4-8(6-9)5-3-7/h2-5,9H,6H2,1H3

InChI key

KMTDMTZBNYGUNX-UHFFFAOYSA-N

General description

4-Methylbenzyl alcohol is widely used in flavoring and fragrances. Used as a precursor to synthesize aldehydes and polycarbonates.

Application

4-Methylbenzyl alcohol was used in the preparation of polycarbonates. It was also used to synthesize alcohols.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Koichiro Mikami et al.
Journal of the American Chemical Society, 135(18), 6826-6829 (2013-05-01)
An organocatalyzed ring-opening polymerization methodology was developed for the preparation of polycarbonates derived from glucose as a natural product starting material. The cyclic 4,6-carbonate monomer of glucose having the 1, 2, and 3 positions methyl-protected was prepared in three steps
Hironao Sajiki et al.
Chemical & pharmaceutical bulletin, 51(3), 320-324 (2003-03-04)
Employment of a Pd/C-pyridine combination as a catalyst is a very useful method for the selective removal (hydrogenolysis) of phenolic O-benzyl, N-Cbz and benzyl ester protective groups and for the selective hydrogenation of nitro and olefin functions of phenol derivatives
Bryce V Plapp et al.
Archives of biochemistry and biophysics, 701, 108825-108825 (2021-03-07)
Enzymes typically have high specificity for their substrates, but the structures of substrates and products differ, and multiple modes of binding are observed. In this study, high resolution X-ray crystallography of complexes with NADH and alcohols show alternative modes of
Carla Villa et al.
International journal of cosmetic science, 30(2), 139-144 (2008-04-02)
Several alcohols--interesting as cosmetic fragrances whose main preparative route on an industrial scale or in the research laboratory is the reduction of the corresponding carbonyl compound--were obtained by a solvent-free methodology in a green chemistry context. The process involves the
Selective Oxidation of Benzyl alcohols to Benzaldyhydes under Phase Transfer Catalysis
Chemical Science Review and Letters, 3, 123-126 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service