Key Documents
HS15994R
Premium Slide box
100 Place, foam lining, white
Synonym(s):
Schiebekasten, slide box microscope, slide storage, slide storage box
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
material
nickel closure
white
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994R
L × W × H
208 × 175 × 34 mm
General description
Slide boxes are perfect for storage or transportation of standard size slides ( 1 x 3in.).
Features and Benefits
  • molded, rigid boxes securely hold slides in grooved slots
  • durable ABS plastic boxes are available wth either cork or foam lining
  • both the lining and the slots are numbered to correspond to the slide inventory sheet
  • closes securely with a rust-resistant, nickel-plated clasp and pin hinges
Choose from one of the most recent versions:
Certificates of Analysis (COA)