Skip to Content
MilliporeSigma
All Photos(1)

Documents

JKA4105

Sigma-Aldrich

HS-PEG1500-SH

average MN 1,500, cross-linking reagent thiol reactive

Synonym(s):

HS-PEG-SH, Polyethylene glycol, Thiol-PEG-Thiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2O(CH2CH2O)nCH2CH2SH
UNSPSC Code:
12162002
NACRES:
NA.23

product name

HS-PEG1500-SH, average Mn 1,500

form

liquid

mol wt

average Mn 1,500

reaction suitability

reagent type: cross-linking reagent
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

−20°C

General description

Thiol PEG Thiol. Thiol reactive PEG crosslinker, reacts with HS groups on cysteine side chains under mild reaction conditions.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service