Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565725

Sigma-Aldrich

Poly(ethylene oxide)

average MN 10,000, 4-arm, thiol terminated, thiol reactive, >40 ppm DTT as stabilizer

Synonym(s):

Polyethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene oxide), 4-arm, thiol terminated, average Mn 10,000, contains >40 ppm DTT as stabilizer

form

solid

mol wt

average Mn 10,000

contains

>40 ppm DTT as stabilizer

reaction suitability

reactivity: thiol reactive

mp

51-56 °C (lit.)

Mw/Mn

<1.2

polymer architecture

shape: 4-arm
functionality: homofunctional

storage temp.

2-8°C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service