Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11124

Sigma-Aldrich

O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol

5000, chemical modification reagent thiol reactive, methoxy, thiol

Synonym(s):

Polyethylene glycol, PEG-thiol, ‘Mercaptopolyethylene glycol monomethyl ether’

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol, 5,000

reaction suitability

reagent type: chemical modification reagent
reactivity: thiol reactive

Ω-end

thiol

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

SMILES string

SCCOCCO.O(CCO)C

InChI

1S/C4H10O2S.C3H8O2/c5-1-2-6-3-4-7;1-5-3-2-4/h5,7H,1-4H2;4H,2-3H2,1H3

InChI key

PVYJAAVBEMPGMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Lanzagorta Lanzagorta Garcia et al.
Materials (Basel, Switzerland), 14(5) (2021-03-07)
An innovative antimicrobial technology for plastic surfaces is presented. We report the synthesis and scale-up of triangular silver nanoplates (TSNPs) and their integration into polycaprolactone (PCL) and polylactic acid (PLA) polymers through a solvent-casting technique. The TSNPs have a high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service