Skip to Content
MilliporeSigma
All Photos(1)

Documents

JKA4095

Sigma-Aldrich

Alkyne-PEG5K

average Mn 5,000, alkyne

Synonym(s):

Alkyne-PEG-Alkyne, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCCCH2NHCOCH2O(CH2CH2O)nCH2CH2OCH2CONHCH2CCH
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Alkyne-PEG5K-Alkyne, average Mn 5,000

form

solid

mol wt

average Mn 5,000

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: homobifunctional

General description

Alkyne-PEG-Alkyne. Click PEG crosslinking reagent for reaction with azide groups.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service