764167
Alkyne-PEG5-acid
Synonym(s):
Polyethylene glycol, Acetylene-PEG5-acid
About This Item
Recommended Products
form
liquid
Quality Level
reaction suitability
reaction type: click chemistry
reagent type: cross-linking reagent
functional group
alkyne
carboxylic acid
storage temp.
−20°C
SMILES string
C#CCOCCOCCOCCOCCOCCC(O)=O
InChI
1S/C14H24O7/c1-2-4-17-6-8-19-10-12-21-13-11-20-9-7-18-5-3-14(15)16/h1H,3-13H2,(H,15,16)
InChI key
GWIACWQVTBMVEI-UHFFFAOYSA-N
Related Categories
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service