Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA3177

Sigma-Aldrich

mPEG5K

average Mn 5,000, Alkyne

Synonym(s):

mPEG-Alkyne, Methoxy-PEG-Alkyne, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2(OCH2CH2)nOCH2CONHCH2CCH
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

mPEG5K-Alkyne, average Mn 5,000

form

solid

mol wt

average Mn 5,000

reaction suitability

reagent type: chemical modification reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Methoxy PEG Alkyne. Click chemistry PEG reagent for reaction with azide groups.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service