Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA10001

Sigma-Aldrich

8arm-PEG10K

average MN 10,000, NH2, tripentaerythritol core, HCl Salt, carboxyl reactive

Synonym(s):

8arm-PEG-NH2, tripentaerythritol core, 8arm-PEG-NH2, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
R(O(CH2CH2O)nCH2CH2NH2HCl)8
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

8arm-PEG10K-NH2, tripentaerythritol core, HCl Salt, average Mn 10,000

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: carboxyl reactive

polymer architecture

shape: 8-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

8arm PEG Amine. Crosslinks into hydrogels. The amine group reacts with carboxylic group (-COOH) or other amine reactive chemical groups. 8ARM(TP)-PEG with tripentaerythritol core has a lower polydispersity and higher molecular weight accuracy compared with the generic 8ARM-PEG with a hexaglycerin core.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Frith, J.E., et al.
Biomaterials, 35(4) (2014)

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service