Skip to Content
MilliporeSigma

406708

Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether

average Mn 440

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C2H5C[CH2[OCH2CH(CH3)]nNH2]3
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
EC Number:
500-105-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

mol wt

average Mn 440

refractive index

n20/D 1.461 (lit.)

density

0.981 g/mL at 25 °C (lit.)

SMILES string

CC(N)CO.CC(O)CO.CCC(CO)(CO)CO

InChI

1S/C6H14O3.C3H9NO.C3H8O2/c1-2-6(3-7,4-8)5-9;1-3(4)2-5;1-3(5)2-4/h7-9H,2-5H2,1H3;3,5H,2,4H2,1H3;3-5H,2H2,1H3

InChI key

MKZVOAOIODOCTL-UHFFFAOYSA-N

General description

Trimethylolpropane tris[poly(propylene glycol), amine terminated] ether (T-403) is a type of polyetheramine that has a primary amine group. It can be used as an epoxy curing agent. T-403 has a hydroxyl group that reacts with the epoxy monomer to form stable ether linkages.

Application

T-403 may be surface functionalized by graphene nanoplatelet (GNP) and can be used as a water coolant in closed conduit flow. It can be incorporated into poly(butylene terephthalate) (PBT) as a blend, which enhances its melt-flowability. It can also be used as a cross-linker, which facilitates the curing of ethylene-glycidyl methacrylate copolymer.


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

235.4 °F - closed cup

Flash Point (°C)

113 °C - closed cup

PPE (personal protective equipment)

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library