Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14526

Sigma-Aldrich

O,O′-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol

Mr 600

Synonym(s):

Jeffamine® ED-600, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

O,O′-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol, 500

description

132 g/eq AHEW (Amine hydrogen equivalent wt)

mol wt

Mr ~600

pH

11.7(lit.)

viscosity

72 cSt(20 °C)(lit.)

mp

-10 °C (lit.)

density

1.035 g/mL (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Flash point 160°C (Lit.)

Application

Modification of polyamides for enhanced hydrophilicity.
Preparation of biocompatible articles and coatings.
Preparation of hydrogels with isocyanates.

Features and Benefits
  • Flexibility from polyether
  • Hydrophilicity from polyethylene glycol.
  • Reactivity of the amine end group.
  • Biocompatibility of polyethylene glycol.

Legal Information

Jeffamine is a registered trademark of Huntsman Petrochemical, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service