Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

D91209

Sigma-Aldrich

Diethyl bromomalonate

92%

Synonym(s):

α-Bromomalonic ester, 1,3-Diethyl 2-bromopropanedioate, 2-Bromomalonic acid diethyl ester, Bromomalonic acid diethyl ester, Diethyl α-bromomalonate, Diethyl 2-bromo-1,3-propanedioate, Diethyl 2-bromomalonate, Diethyl 2-bromopropanedioate, Diethyl bromopropanedioate, Ethyl bromomalonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH(CO2CH2CH3)2
CAS Number:
Molecular Weight:
239.06
Beilstein:
509768
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

92%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

233-235 °C (lit.)

density

1.402 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(Br)C(=O)OCC

InChI

1S/C7H11BrO4/c1-3-11-6(9)5(8)7(10)12-4-2/h5H,3-4H2,1-2H3

InChI key

FNJVDWXUKLTFFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Contains diethyl dibromomalonate, and varying amounts of diethyl malonate

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Kujawa et al.
Polymers, 11(7) (2019-07-25)
1-(1,3-diethoxy-1,3-dioxopropan-2-ylo)-3-methylimidazolium bromide (RIL1_Br), 1-(2-etoxy-2-oxoethyl)-3-methylimidazolium bromide (RIL2_Br), 1-(2-etoxy-2-oxoethyl)-3-methylimidazolium tetrafluoroborate (RIL3_BF4) ionic liquids were synthesized. Subsequently, the dense cellulose acetate propionate (CAP)-based materials containing from 9 to 28.6 wt % of these reactive ionic liquids were elaborated. Reactive ionic liquids (RILs) were
Jinjin Shi et al.
International journal of nanomedicine, 8, 1551-1562 (2013-05-03)
Fullerene (C60) has shown great potential in drug delivery. In this study we exploited modified fullerene (diadduct malonic acid-fullerene-Asn-Gly-Arg peptide [DMA-C60-NGR]) as an antitumor drug carrier in order to build a new tumor-targeting drug delivery system. We also investigated the
Shefang Ye et al.
Nanoscale research letters, 10(1), 953-953 (2015-06-11)
Fullerene (C60) derivatives, a unique class of compounds with potent antioxidant properties, have been reported to exert a wide variety of biological activities including neuroprotective properties. Mitochondrial dynamics are an important constituent of cellular quality control and function, and an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service