Skip to Content
MilliporeSigma
All Photos(1)

Documents

860271

Sigma-Aldrich

L-Valine methyl ester hydrochloride

99%

Synonym(s):

(S)-Valine methyl ester hydrochloride, Methyl (S)-2-amino-3-methylbutanoate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
167.63
Beilstein:
3594960
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

optical activity

[α]/D +15°, c = 2 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

171-173 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl.COC(=O)[C@@H](N)C(C)C

InChI

1S/C6H13NO2.ClH/c1-4(2)5(7)6(8)9-3;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1

InChI key

KUGLDBMQKZTXPW-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beibei Hu et al.
International journal of pharmaceutics, 547(1-2), 637-647 (2018-06-23)
Thermogels, used as multi-functional drug-loading materials, have properties that mainly rely on their gelator structure. Although a large variety of organogel systems are used as drug delivery carriers, relatively few have been investigated in terms of their structure-property correlations based
Sonu Kumar et al.
Macromolecular bioscience, 20(6), e2000048-e2000048 (2020-04-15)
The synthesis and self-assembly of peptide-polymer conjugates into fibrillar nanostructures are reported, based on the amyloidogenic peptide KLVFF. A strategy for rational synthesis of polymer-peptide conjugates is documented via tethering of the amyloidogenic peptide segment LVFF (Aβ17-20 ) and its
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service