Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D84601

Sigma-Aldrich

Diethyl acetamidomalonate

98%

Synonym(s):

Acetamidomalonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH(CO2C2H5)2
CAS Number:
Molecular Weight:
217.22
Beilstein:
783883
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

bp

185 °C/20 mmHg (lit.)

mp

95-98 °C (lit.)

SMILES string

CCOC(=O)C(NC(C)=O)C(=O)OCC

InChI

1S/C9H15NO5/c1-4-14-8(12)7(10-6(3)11)9(13)15-5-2/h7H,4-5H2,1-3H3,(H,10,11)

InChI key

ISOLMABRZPQKOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambasivarao Kotha et al.
Amino acids, 41(4), 933-936 (2010-11-10)
Strategically a new approach for the synthesis tetrahydro-β-carboline unit with the aid of diethyl acetamidomalonate as a glycine equivalent has been described.
Soraia Veloso Silva Santana et al.
Journal of esthetic and restorative dentistry : official publication of the American Academy of Esthetic Dentistry ... [et al.], 21(6), 397-404 (2009-12-17)
Surface sealants may reduce or avoid problems related to the marginal interface. The aim of this study was to evaluate the microleakage in resin composite Class V restorations sealed with an adhesive system (Xeno III [Dentsply, Konstanz, Germany]), a sealant
Zsolt Rapi et al.
Carbohydrate research, 365, 61-68 (2012-12-12)
The synthesis of four new ribo-hexopyranoside-based chiral lariat ethers of monoaza-15-crown-5 type and two altropyranoside-based crown ethers were elaborated. Our syntheses utilized the regioselective ring opening of the oxiran moiety of the 2,3-anhydro sugars by nucleophilic reagents to afford the
G K Powell et al.
Preparative biochemistry, 11(3), 339-350 (1981-01-01)
This paper describes the complete chemical synthesis of 4-methylene-DL-glutamic acid from diethylmalonate, formaldehyde and diethyl acetamidomalonate. The amino acid was obtained pure following ion-exchange chromatography and/or crystallization from hot water in an overall yield of 30% based on the amount

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service