Key Documents
CAD6020
1-way stopcock
female Luer lock to female Luer lock
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 6020
Related Categories
General description
Stopcocks are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Choose from one of the most recent versions:
Certificates of Analysis (COA)