Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7396

Supelco

Liquid chromatography column fittings

Stopcock, Luer Lock (A)

Synonym(s):

polymeric adsorbent, 50-80 mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

grade

for chromatography

technique(s)

LPLC: suitable

Looking for similar products? Visit Product Comparison Guide

General description

A polycarbonate/acetal copolymer on-off control with female Luer-Lock inlet and male Luer-Lock outlet ports.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Antibody purification protocols yield preparations containing endogenous IgG alongside specific antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service