Documents
20926
PTFE Body Two-Way Valve
for use with Hamilton PTFE Luer Lock (TLL) and other Luer lock syringes.
About This Item
UNSPSC Code:
41122001
material
PTFE valve body
feature
positions 2
compatibility
for use with Hamilton PTFE Luer Lock (TLL) and other Luer lock syringes.
General description
This miniature, two-way valve from Hamilton provides flexibility, inertness, and small size for collecting gas samples and transporting or storing them in the original sampling syringe.
Fluid path is made entirely of PTFE to ensure inertness. Can be autoclaved for applications requiring sterility. 100 psig maximum pressure. Fits Hamilton PTFE Luer lock (TLL) and other Luer lock syringes.
  • Lever-actuated on/off valve for storing and transporting samples.
  • KEL-F hub needles attach to the valve
  • Bore ID 0.39"
Application
PTFE body two-way valve has been used in instrumental analysis of Cloud-point polyethylene glycol for imaging of immobilised single molecules using total-internal-reflection microscopy.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Customers Also Viewed
Cloud-point PEG Glass Surfaces for Imaging of Immobilized Single Molecules by Total-internal-reflection Microscopy
Zhang, Zhengjian, et al.
Bio-protocol, 6, 2331-8325 (2016)