Key Documents
CAD6011
3-way stopcock
female Luer lock to male Luer lock, right side, female Luer lock, compression-nut
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 6011
General description
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
Choose from one of the most recent versions:
Certificates of Analysis (COA)