Key Documents
BR32808
BRAND® USP BLAUBRAND® class A measuring cylinder, tall form
volume 10 mL
Synonym(s):
BLAUBRAND®, Graduated Cylinders, USP
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
borosilicate glass
feature
DE-M marking
manufacturer/tradename
BRAND 32808
volume
10 mL
General description
The BRAND® USP BLAUBRAND® class A measuring cylinder in tall form is crafted from high-quality Boro 3.3 glass, ensuring durability and precision. It complies with DIN EN ISO 4788 standards and is calibrated to contain (TC, In), making it suitable for accurate liquid measurement. The design features a spout for easy pouring and a hexagonal glass base for stability. Each measuring cylinder comes with a USP batch certificate. This measuring cylinder is an essential tool for laboratories requiring high accuracy and reliability in liquid measurement.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)