Key Documents
BR23836
BRAND® spare burette length for compact automatic burette
25 mL, SILBERBRAND
Synonym(s):
Accessories, Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
clear borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
BRAND 23836
measuring range
25 mL
burette L
425 mm
volume
25 mL
General description
The BRAND® spare burette length for the compact automatic burette is made from high-quality Boro 3.3 clear glass, ensuring durability and clarity for precise measurements. Designed specifically for compatibility with BLAUBRAND® and SILBERBRAND compact automatic burettes, this spare part maintains the performance standards required for accurate liquid dispensing.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG