Key Documents
BR13943
BRAND® spare burette tubes for compact burette
size 10 mL, BLAUBRAND®, clear borosilicate glass 3.3
Synonym(s):
Accessories, BLAUBRAND®, Burettes, Compact Burettes
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
clear borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
BRAND 13943
L
700 mm
size
10 mL
Related Categories
General description
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)