Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BMSE1101SK

Benchmark myGel Mini Electrophoresis System Starter Kit

AC/DC input 115 V

Synonym(s):

protein separation, gel catsing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.13

manufacturer/tradename

Benchmark Scientific E1101-SK

Looking for similar products? Visit Product Comparison Guide

General description

myGel Mini Electrophoresis Starter kit includes mini gel box with integrated power supply, casting set, small bottle of agarose and a mini balance

Application

Electrophoresis

Legal Information

myGel is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service