Key Documents
BMSC3100
Benchmark LC-8 3500 Centrifuge with 8 x 15ml rotor
speed 400-3,500 rpm, AC/DC input 120 V
Synonym(s):
laboratory centrifuge, benchtop centrifuge, bendh top centrifuge, lab centrifuge
About This Item
UNSPSC Code:
40161701
NACRES:
NB.11
feature
refrigerated: no
to hold 8 x 10 or 15 mL tubes
manufacturer/tradename
Benchmark Scientific C3100
parameter
400-3,500 rpm speed
General description
Benchmark′s LC-8 series of laboratory centrifuges is uniquely designed to meet a wide range of applications in both the clinical and research laboratory. An eight position rotor is included and designed to directly accept 10ml and 15ml test-tubes (conical or round bottom), while optional adapters are also available for use with all popular sizes of blood collection and culture tubes. Unlike most compact centrifuges in this class, the rotor can be easily removed from the chamber for cleaning.
The back-lit, liquid crystal display provides full control over the set run time and rotational speed. The rotational speed can be set/viewed in rpm or rcf and acceleration/deceleration levels can be easily adjusted in the advanced settings mode.
To ensure safety, the digital microprocessor ensures that the lid has been closed and that the safety lid lock is engaged prior to starting a centrifugation cycle. In addition, an advanced imbalance detection system constantly monitors the balance and vibration, automatically bringing the rotor to a stop if an imbalance is detected.