Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BAH372600000

SP Bel-Art® Liquid Nitrogen Cooled Mortar with handle

stainless steel (with polyethylene housing), diam. × H 16.5 cm (6.5 in.) × 11.4 cm (4.5 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43
Pricing and availability is not currently available.

material

stainless steel (with polyethylene housing)

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H37260-0000

diam. × H

16.5 cm (6.5 in.) × 11.4 cm (4.5 in.)

diam.

6.5 in.

Related Categories

General description

Useful for grinding frozen samples, this product includes a heavily insulated blue polyethylene housing that keeps liquid nitrogen cold longer.

Features and Benefits

  • 304 stainless steel mortar suspended above a stainless steel bowl holds liquid nitrogen
  • Spare mortars are useful for grinding different types of samples on the same day
  • Insulated handle is removable for saving space; 10.2cm (4”) long
  • Three suction cup feet stabilize the unit on the workbench
  • Recommended size pestle is any size appropriate for a 275ml mortar (not included)

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service