Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z370517

Nalgene® Labtop cooler

0°C Labtop cooler, clear lid

Synonym(s):

PCR amplification cooler, PCR amplification workstation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025
NACRES:
NB.22

packaging

pack of 1 ea

manufacturer/tradename

Nalgene® 5116-1600

L × W × H

197 mm × 140 mm × 190 mm

weight

3.5 lb

Looking for similar products? Visit Product Comparison Guide

General description

Maintains twelve 16-17 mm diameter tubes below 2°C for up to 4 hrs at room temperature.
Molded from durable polycarbonate and filled with non-toxic freezable gel. A convenient alternative to ice buckets for keeping reagents and enzymes cool. Also protects critical samples from temperature fluctuations in the freezer.

Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service