Key Documents
BAF134460000
SP Bel-Art® I.D. Rack Tags
polypropylene, width × H × L 38 mm (1.5 in.) × 0.125 in. (3 mm) × 1.5 in. (89 mm)
Synonym(s):
Wire Rack I.D. Tags
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F13446-0000
width × H × L
38 mm (1.5 in.) × 0.125 in. (3 mm) × 1.5 in. (89 mm)
General description
Durable, reusable and easy to attach, Scienceware® Wire Rack I.D. Tags eliminate the challenges of labeling all wire racks, trays, baskets and other wire-formed items made of 3mm (1/8") or 5mm (3/16") wire.
Features and Benefits
  • Smooth, polypropylene plastic surface allows labeling directly onto the tag with permanent marker, dry-erase marker or a pressure sensitive adhesive label
  • White color provides excellent contrast for most ink colors
  • One piece, write on, wipe off design is economical, easy to use and does not require assembly
  • Chemical resistant and autoclavable at 121°C (250°F)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)