Skip to Content
MilliporeSigma
All Photos(1)

Documents

B56498

Sigma-Aldrich

3-Bromoanisole

≥98%

Synonym(s):

1-Bromo-3-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCH3
CAS Number:
Molecular Weight:
187.03
Beilstein:
1524687
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

refractive index

n20/D 1.563 (lit.)

bp

210-211 °C (lit.)

density

1.477 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(Br)c1

InChI

1S/C7H7BrO/c1-9-7-4-2-3-6(8)5-7/h2-5H,1H3

InChI key

PLDWAJLZAAHOGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F

Flash Point(C)

95 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoyuki Yasaka et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(10), 1183-1188 (2018-10-12)
Aryl halides are a very important category of compounds that include many vital drugs and key industrial additives, such as clofibrate and bromobenzene, respectively. Due to their importance, our research group previously developed a novel fluorescence labeling approach for their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service