Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

439479

Sigma-Aldrich

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

greener alternative

>95% in F+ active

Synonym(s):

N-Chloromethyl-N′-fluorotriethylenediammonium bis(tetrafluoroborate), F-TEDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14B2ClF9N2
CAS Number:
Molecular Weight:
354.26
Beilstein:
5368649
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reagent type: catalyst
reagent type: oxidant
reaction type: C-H Activation

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

>95% in F+ active

mp

260 °C (lit.)

functional group

chloro

greener alternative category

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2

InChI

1S/C7H14ClFN2.2BF4/c8-7-10-1-4-11(9,5-2-10)6-3-10;2*2-1(3,4)5/h1-7H2;;/q+2;2*-1

InChI key

TXRPHPUGYLSHCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is a fluorine donor also called Selectfluor and has been enhanced for catalysis. Find details here.

Application

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) is an electrophilic fluorinating reagent used for greener fluorination of acetoacetamides in PEG-400.
Catalyst-free and highly selective electrophilic mono-fluorination of acetoacetamides: facile and efficient preparation of 2-fluoroacetoacetamides in PEG-400

It can be used:
  • As a highly effective and versatile source of electrophilic fluorine.
  • As a fluorinating reagent that effects oxidative ring opening of 2,5-diarylfurans to cis-1,4-enediones.
  • For direct fluorination of silyl ketene acetals leading to the formation of α-fluoro-α-arylcarboxylic acids.
  • As an electrophilic fluorinating reagent used. e.g. in the preparation of fluorinated acylsilanes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajendra P Singh et al.
Accounts of chemical research, 37(1), 31-44 (2004-01-21)
Synthetic and structural aspects of organofluorine compounds continue to be the focal points of vigorous research activities, as evidenced by the appearance of a large number of publications. Among the various useful methodologies for the introduction of fluorine into organic
Oxidative ring opening of 2, 5-diarylfurans by Selectfluor
Blank SJ and Stephens CE
Tetrahedron Letters, 47(38), 6849-6850 (2006)
A general method for synthesis of trifluoroacetyltrialkyl (aryl) silanes and the Sakurai reaction of fluorinated acylsilanes with allyl silanes
Chung WJ and Welch JT
Journal of Fluorine Chemistry, 125(4), 543-548 (2004)
Paul T Nyffeler et al.
Angewandte Chemie (International ed. in English), 44(2), 192-212 (2004-12-04)
The replacement of hydrogen atoms with fluorine substituents in organic substrates is of great interest in synthetic chemistry because of the strong electronegativity of fluorine and relatively small steric footprint of fluorine atoms. Many sources of nucleophilic fluorine are available
A novel general method for preparation of a-fluoro-a-arylcarboxylic acid. Direct fluorination of silyl ketene acetals with Selectfluor
Zhang F and Song JZ
Tetrahedron Letters, 47(43), 7641-7644 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service