Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

B22003

Sigma-Aldrich

2-Benzylaminoethanol

95%

Synonym(s):

N-Benzylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHCH2CH2OH
CAS Number:
Molecular Weight:
151.21
Beilstein:
775164
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.543 (lit.)

bp

153-156 °C/12 mmHg (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

OCCNCc1ccccc1

InChI

1S/C9H13NO/c11-7-6-10-8-9-4-2-1-3-5-9/h1-5,10-11H,6-8H2

InChI key

XNIOWJUQPMKCIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of formaldehyde in air as an oxazolidine derivative by capillary gas chromatography.
E R Kennedy et al.
Analytical chemistry, 54(11), 1739-1742 (1982-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service