Skip to Content
MilliporeSigma
All Photos(1)

Documents

130001

Sigma-Aldrich

1-Methylpiperazine

99%

Synonym(s):

N-Methylpiperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2
CAS Number:
Molecular Weight:
100.16
Beilstein:
102724
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.5 (vs air)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

138 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

CN1CCNCC1

InChI

1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3

InChI key

PVOAHINGSUIXLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Methylpiperazine is a heterocyclic diamine used as a versatile building block and intermediate in organic synthesis.

Application

1-Methylpiperazine was used as mimic template in the preparation of molecularly imprinted microspheres (MIMs). It was also used to prepare the difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

88.7 °F - closed cup

Flash Point(C)

31.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juxiang Ruan et al.
Journal of chromatography. A, 1297, 77-84 (2013-06-01)
This paper reports the preparation of a novel, silica-based, strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative. To prepare the difunctional strong anion-exchange stationary phase, activated silica beads were first bonded with 3-chloropropyltriethoxysilane and then reacted with 1-methylpiperazine followed by
Hongyuan Yan et al.
The Analyst, 137(12), 2884-2890 (2012-05-04)
A highly selective molecularly imprinted solid-phase extraction (MISPE) combined with liquid chromatography-ultraviolet detection was developed for the simultaneous isolation and determination of four plant hormones including indole-3-acetic acid (IAA), indole-3-propionic acid (IPA), indole-3-butyric acid (IBA) and 1-naphthaleneacetic acid (NAA) in
E Gavathiotis et al.
Nucleic acids research, 28(3), 728-735 (2000-01-19)
The solution structure of the dodecamer duplex d(CTTTTGCAAAAG)(2)and its 2:1 complex with the bis -benzimidazole Hoechst 33258 has been investigated by NMR and NOE-restrained molecular dynamics (rMD) simulations. Drug molecules are bound in each of the two A-tracts with the
Olesya A Troshina et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(21), 5569-5577 (2006-06-07)
An oxidative radical photoaddition of mono N-substituted piperazines to [60]fullerene was systematically investigated. Reactions of C60 with piperazines bearing bulky electron-withdrawing groups (2-pyridyl, 2-pyrimidinyl) were found to be the most selective and yielded C60(amine)4O as major products along with small
Andrius Žilionis
European journal of mass spectrometry (Chichester, England), 26(1), 25-35 (2019-08-21)
Liquid chromatography-tandem mass spectrometry is one of the most sensitive tools for determination of trace amounts of analytes in metabolomics and proteomics. The highest sensitivity is achieved in selected reaction monitoring detection, which involves fragmentation of the molecular ion between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service