Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00612

(S)-N-(1-(Dimethylamino)-3-phenylpropan-2-yl)acetamide

≥95%

Synonym(s):

Yu MPAAM ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O
CAS Number:
Molecular Weight:
220.31
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

Application

(S)-N-(1-(Dimethylamino)-3-phenylpropan-2-yl)acetamide is an MPAAM ligand developed in the Yu Group for the Pd-catalyzed enantioselective C(sp3)-H arylation of fre cyclopropanecarboxylic and aminoisobutyric acids.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng-Xiang Shen et al.
Journal of the American Chemical Society, 140(21), 6545-6549 (2018-05-10)
A monoprotected aminoethyl amine chiral ligand based on an ethylenediamine backbone was developed to achieve Pd-catalyzed enantioselective C(sp3)-H arylation of cyclopropanecarboxylic and 2-aminoisobutyric acids without using exogenous directing groups. This new chiral catalyst affords new disconnection for preparing diverse chiral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service