Skip to Content
MilliporeSigma
All Photos(2)

Documents

149861

Sigma-Aldrich

2-Furoyl chloride

95%

Synonym(s):

Furan-2-carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClO2
CAS Number:
Molecular Weight:
130.53
Beilstein:
110144
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.531 (lit.)

bp

173-174 °C (lit.)

mp

−2 °C (lit.)

density

1.324 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccco1

InChI

1S/C5H3ClO2/c6-5(7)4-2-1-3-8-4/h1-3H

InChI key

OFTKFKYVSBNYEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Furoyl chloride is used in the chararcterisation of mixtures of polyethoxylated alcohols and their sulphates. It was also used in the preparation of S-2-furoyl-O-alkyldithiocarbonic anhydrides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Morvan et al.
Analytical and bioanalytical chemistry, 384(6), 1409-1415 (2006-02-28)
Bachus et al. [1] recently described a new derivatisation method using 2-furoyl chloride for the characterisation of mixtures of polyethoxylated alcohols and their corresponding sulfates. This paper deals with the control of the derivatisation steps; hydrolysis and extraction conditions were
Ethel A Wilhelm et al.
Canadian journal of physiology and pharmacology, 98(5), 304-313 (2019-12-11)
The present study evaluated the in vitro acetylcholinesterase (AChE) inhibitor activity of two new selanyl amide derivatives in cerebral structures of mice. Our results demonstrated that N-(2-(3-(phenylselanyl)propoxy)phenyl)furan-2-carboxamide (1) and N-(2-(3-(phenylselanyl)propoxy)phenyl)thiophene-2-carboxamide (2) inhibited the in vitro AChE activity in mice. Another
J Tułecki et al.
Polish journal of pharmacology and pharmacy, 37(5), 709-715 (1985-09-01)
New S-2-furoyl-O-alkyldithiocarbonic anhydrides have been obtained by condensation of 2-furoyl chloride with potassium O-alkyl-dithiocarbonates. The toxicity and radiosensitizing activity of these compounds were determined.
Christoph Cerny et al.
Food chemistry, 337, 128008-128008 (2020-09-14)
This study investigated the role of furfuryl alcohol (FFA) in the formation of furfurylthiol (FFT), the most important odorant in roasted coffee, using in-bean and spiking experiments. Green beans were spiked with FFA, and after roasting FFT was quantified by
Won-Ji Lee et al.
Polymers, 12(3) (2020-03-12)
In this paper, a self-healable nanocomposite based on the Diels-Alder reaction is developed. A graphene-based nanofiller is introduced to improve the self-healing efficiency, as well as the mechanical properties of the nanocomposite. Graphene oxide (GO) is modified with maleimide functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service