Skip to Content
MilliporeSigma
All Photos(2)

Documents

P62208

Sigma-Aldrich

3-Pyridinecarboxaldehyde

98%

Synonym(s):

Nicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO
CAS Number:
Molecular Weight:
107.11
Beilstein:
105343
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.549 (lit.)

bp

78-81 °C/10 mmHg (lit.)

density

1.141 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cccnc1

InChI

1S/C6H5NO/c8-5-6-2-1-3-7-4-6/h1-5H

InChI key

QJZUKDFHGGYHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Narender et al.
Bioorganic & medicinal chemistry, 14(13), 4600-4609 (2006-03-03)
Baylis-Hillman acetates were synthesized from substituted 2-chloronicotinaldehydes and were conveniently transformed into multisubstituted quinolines and cyclopenta[g]quinolines on reaction with nitroethane or ethyl cyanoacetate via a successive S(N)2'-S(N)Ar elimination strategy. Thus, synthesized quinolines were evaluated for antimicrobial activity and found having
C C Shone et al.
Biochemistry, 20(26), 7494-7501 (1981-12-22)
Coenzyme A linked aldehyde dehydrogenase from Escherichia coli strain B has been purified to a specific activity of 14 units/mg of protein, and initial rate and substrate analogue inhibition experiments have been performed. On the basis of these steady-state measurements
[Selenazoles. IX. [9]. 3-methyl-4-R-selenazolidene-2-hydrazones of acetaldehyde, benzaldehyde, isonicotinaldehyde, nicotinaldehyde and picolinaldehyde].
S Biliński et al.
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina, 39, 25-31 (1984-01-01)
P Narender et al.
Bioorganic & medicinal chemistry letters, 15(24), 5378-5381 (2005-10-11)
New Baylis-Hillman adducts are synthesized based on substituted 2-chloronicotinaldehydes and screened for their in vitro anti-malarial activity against chloroquine sensitive and chloroquine resistant Plasmodium falciparum. Out of the six new compounds synthesized and screened, 2b, 2c and 2d compounds showed
K S De Jongh et al.
The Biochemical journal, 242(1), 143-150 (1987-02-15)
The kinetic mechanism of the major sheep liver aldehyde reductase (ALR1) was studied with three aldehyde substrates: p-nitrobenzaldehyde, pyridine-3-aldehyde and D-glucuronate. In each case the enzyme mechanism was sequential and product-inhibition studies were consistent with an ordered Bi Bi mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service