Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A42050

Sigma-Aldrich

4-Aminobenzyl cyanide

99%

Synonym(s):

4-Aminophenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2CN
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

lumps

bp

312 °C (lit.)

mp

45-48 °C (lit.)

SMILES string

Nc1ccc(CC#N)cc1

InChI

1S/C8H8N2/c9-6-5-7-1-3-8(10)4-2-7/h1-4H,5,10H2

InChI key

YCWRFIYBUQBHJI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service