Skip to Content
MilliporeSigma
All Photos(1)

Documents

C89609

Sigma-Aldrich

4-Formylbenzonitrile

95%

Synonym(s):

4-Cyanobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4CHO
CAS Number:
Molecular Weight:
131.13
Beilstein:
606473
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

bp

133 °C/12 mmHg (lit.)

mp

100-102 °C (lit.)

SMILES string

O=Cc1ccc(cc1)C#N

InChI

1S/C8H5NO/c9-5-7-1-3-8(6-10)4-2-7/h1-4,6H

InChI key

WZWIQYMTQZCSKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Formylbenzonitrile can be used in the synthesis of various boron-dipyrromethenes (BODIPY), porphyrins, corroles and other related macrocycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rothemund and Adler-Longo reactions revisited: synthesis of tetraphenylporphyrins under equilibrium conditions.
Lindsey J S, et al.
The Journal of Organic Chemistry, 52(5), 827-836 (1987)
meso-Pyrimidinyl-substituted A2B-and A3-corroles.
Ngo T H, et al.
The Journal of Organic Chemistry, 75(6), 2127-2130 (2010)
Unusually High Performance Photovoltaic Cell Based on a [60] Fullerene Metal Cluster? Porphyrin Dyad SAM on an ITO Electrode.
Cho Y J, et al.
Journal of the American Chemical Society, 127(8), 2380-2381 (2005)
Accessing near-infrared-absorbing BF2-azadipyrromethenes via a push?pull effect.
Jiao L, et al.
The Journal of Organic Chemistry, 79(4), 1830-1835 (2014)
Minna K Rahnasto et al.
Bioorganic & medicinal chemistry, 19(23), 7186-7193 (2011-10-25)
The human CYP2A6 enzyme metabolises several xenobiotics including nicotine, the addictive component in tobacco. Reduced activity of CYP2A6, either for genetic reasons or by administering inhibitors of CYP2A6, reduces tobacco smoking. The aim was to design novel inhibitors of CYP2A6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service