Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

169986

Sigma-Aldrich

4-Methoxyphenylacetonitrile

97%

Synonym(s):

4-Methoxybenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CN
CAS Number:
Molecular Weight:
147.17
Beilstein:
509162
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.531 (lit.)

bp

286-287 °C (lit.)

density

1.085 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CC#N)cc1

InChI

1S/C9H9NO/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5H,6H2,1H3

InChI key

PACGLQCRGWFBJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methoxyphenylacetonitrile has been isolated from the sponge Psammaplysilla purpurea.

Application

4-Methoxyphenylacetonitrile was used as starting reagent in the synthesis of :
  • 2-(1-cyano-1-(2-methoxy)phenyl)methylidene-3-phenylthiazolidine-4,5-dione
  • 2-(1-cyano-1-(4-methoxy)-phenyl)methylidene-3-phenylthiazolidine-4,5-dione
  • α-cyanostilbenes

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

242.6 °F - DIN 51758

Flash Point(C)

117 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brominated benzeneacetonitriles, the dibromotyrosine metabolites from the sponge Psammaplysilla purpurea.
Venkateswarlu Y and Chavakula R.
Journal of Natural Products, 58(7), 1087-1088 (1995)
Uwe Albrecht et al.
Bioorganic & medicinal chemistry, 13(14), 4402-4407 (2005-05-21)
2-Alkylidenethiazolidine-4,5-diones were prepared by novel one-pot cyclizations of arylacetonitriles with isothiocyanates and ethyl 2-chloro-2-oxoacetate. The products show antibiotic activity against the Gram-positive bacteria Bacillus subtilis and Staphylococcus aureus.
Kavya S Keremane et al.
Photochemistry and photobiology, 97(2), 289-300 (2020-10-02)
Developing effective and low-cost organic hole-transporting materials (HTMs) is crucial for the construction of high-performance perovskite solar cells (PSCs) and to promote their production in commercial ventures. In this context, we herein report the molecular design, synthesis and characterization of
Byung Seok Moon et al.
Bioorganic & medicinal chemistry, 17(9), 3479-3488 (2009-04-11)
We have investigated halogen-substituted non-steroidal estrogens with selective binding affinity for the estrogen receptor beta (ERbeta that might be used for imaging the levels of this ER-subtype in breast tumors by positron emission tomography (PET). Based on diarylpropionitrile (DPN, 1a)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service