Skip to Content
MilliporeSigma
All Photos(1)

Documents

933376

Sigma-Aldrich

SEM Sample Holder

SEM Pin Stub Adapter, size 12.5 mm

Synonym(s):

stub adapter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

size

12.5 mm

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

A simple to use adapter which will convert Hitachi, JEOL or ISI microscopes to use ⅛" pin stubs or Cambridge stubs.
Made of aluminum with spring retainers which will give a good connection.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service