Skip to Content
MilliporeSigma

C9802

Cover glasses

size 22 mm × 22 mm

Synonym(s):

Glass Cover Slips, Microscope Cover Glass

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
MA.01
UNSPSC Code:
41122600
Packaging:
case of 10 1-oz pkgs, pkg of ~155 per ounce
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

No. 1 thickness (0.13-0.16 mm)

packaging

case of 10 1-oz pkgs, pkg of ~155 per ounce

size

22 mm × 22 mm

Application

Cover glasses have been used:
  • to mount the tissue sections on the slide for confocal laser scanning microscopic analysis
  • to scrape off excess mucus and residual feces from intestinal tissue during organoids preparation
  • in the microinjection chamber



Still not finding the right product?

Explore all of our products under Cover glasses




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library