Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

932132

Sigma-Aldrich

2PXZ-OXD

≥99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H24N4O3
CAS Number:
Molecular Weight:
584.62
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
Assay:
≥99% (HPLC)
grade:
sublimed grade
solubility:
toluene: soluble

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

description

3.2 eV (Eg: HOMO-LUMO gap)

loss

0.5% TGA, > 250 °C (weight loss)

solubility

toluene: soluble

fluorescence

λem 512 nm±10 nm in toluene

Orbital energy

HOMO -5.3 eV 
LUMO -2.1 eV 

λ

291 nM in toluene
in toluene

UV absorption

λ: 291 nm±5 nm Amax

Application

2PXZ-OXD can be employed in organic light-emitting diodes (OLEDs) as an electron-transport layer, facilitating the efficient injection and transport of negative charges (electrons) from the cathode to the emissive layer. 2PXZ-OXD exhibits good electron mobility and excellent electron-blocking properties, contributing to improved charge transport and overall device performance in OLEDs. Furthermore, 2PXZ-OXD can be utilized in studies such as organic photovoltaic (OPV) devices and organic field-effect transistors (OFETs) due to its electron transport and blocking characteristics.
Use as TADF Green Dopant, with a EQE of 14.9%.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxadiazole- and triazole-based highly-efficient thermally activated delayed fluorescence emitters for organic light-emitting diodes.
Lee J, et al.
Journal of Material Chemistry C, 1, 4599-4604 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service