Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

900940

Sigma-Aldrich

DMAC-DPS

sublimed

Synonym(s):

10,10′-(4,4′-Sulfonylbis(4,1-phenylene))bis(9,9-dimethyl-9,10-dihydroacridine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H36N2O2S
CAS Number:
Molecular Weight:
632.81
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

description

Absorption (UV): 286 nm (in toluene)
Emission (PL): 469 nm (in toluene)

Quality Level

Assay

≥99% (Sublimed, HPLC)

form

solid

Looking for similar products? Visit Product Comparison Guide

General description

Thermally activated delayed fluorescent (TADF) blue dopant.

Application

  • Diluted exciplex concentrations in organic light emitting diodes for blue-shifted spectra and improved efficiency: This study explores the use of DMAC-DPS and PO-T2T to achieve blue-shifted spectra and improved efficiency in OLEDs (J Yan, et al., 2022).
  • Non-doped white organic light-emitting devices consisting of thermally activated delayed fluorescent emitters and ultrathin phosphorescent emitters: The research focuses on using DMAC-DPS as an emitting layer for high quantum efficiency in non-doped white OLEDs (Q Li, et al., 2019).
  • The optical spectra of DMAC‐based molecules for organic light‐emitting diodes: A density functional theory study of DMAC-DPS and other DMAC-based molecules to understand their emission properties (Y Wang, et al., 2022).
  • Highly efficient and bright blue organic light-emitting devices based on solvent engineered, solution-processed thermally activated delayed fluorescent emission layer: The study investigates the use of DMAC-DPS for efficient carrier transport and high brightness in blue OLEDs (J Yang, et al., 2019).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qisheng Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 27(12), 2096-2100 (2015-02-14)
The design of efficient and concentration-insensitive metal-free thermally activateddelayed fluorescence (TADF) materials is reported. Blue and green organic light-emitting diodes (OLEDs) containing a hole-transport layer, an undoped TADF emissive layer, and an electron-transport layer achieve maximum external quantum efficiencies of
Efficient blue organic light-emitting diodes employing thermally activated delayed fluorescence
Zhang et al.
Nature Photonics, 8(4), 326-332 (2014)

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service