Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

931004

Sigma-Aldrich

9-(3′-(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl-3-yl)-9H-carbazole

≥99% (HPLC)

Synonym(s):

Cz-m2Ph-TRZ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H26N4
CAS Number:
Molecular Weight:
550.65
MDL number:
UNSPSC Code:
12352207
NACRES:
NA.23

grade

sublimed grade

Quality Level

description

T1 = 2.81 eV

Assay

≥99% (HPLC)

λmax

280 nm±10 nm in toluene

Orbital energy

HOMO 6.08 eV 
LUMO 3.28 eV 

Application

It′s popularly used in OLEDs as a host TADF material, contributing to increased efficiency (exciplex efficient emitter) and long lifetime OLED device.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Efficiency Exciplex Emitters Using Donor?Acceptor Type Acceptor Material.
Seok O C, et al.
The Journal of Physical Chemistry C, 119(39), 22618?22624-22618?22624 (2015)
Dongdong Zhang et al.
ACS applied materials & interfaces, 8(6), 3825-3832 (2016-01-23)
Exciplex forming cohosts have been widely adopted in phosphorescent organic light-emitting diodes (PHOLEDs), achieving high efficiency with low roll-off and low driving voltage. However, the influence of the exciplex-forming hosts on the lifetimes of the devices, which is one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service