Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

925136

Sigma-Aldrich

4,6-Dimethyl-2-(methylsulfinyl)-3-nitropyridine

Synonym(s):

2-Sulfonylpyridine, Cysteine-reactive electrophile, SNAr probe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N2O3S
CAS Number:
Molecular Weight:
214.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

reaction suitability

reagent type: chemical modification reagent

Quality Level

storage temp.

2-8°C

Application

4,6-Dimethyl-2-(methylsulfinyl)-3-nitropyridine is a sulfoxide-containing fragment for covalent ligand discovery via cysteine labeling as demonstrated by Zamboldo and coworkers. Fragment electrophiles are useful in chemoproteomic and ligandability studies for both traditionally druggable proteins as well as ″undruggable,″ or difficult-to-target, proteins.

Related useful products may include cysteine fragment KB02 (912131), IA alkyne (924237), IA 5-TAMRA (925020), desthiobiotin iodoacetamide (923826), or biotin iodoacetamide (B2059).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio Zambaldo et al.
Journal of the American Chemical Society, 142(19), 8972-8979 (2020-04-18)
The emerging use of covalent ligands as chemical probes and drugs would benefit from an expanded repertoire of cysteine-reactive electrophiles for efficient and diverse targeting of the proteome. Here we use the endogenous electrophile sensor of mammalian cells, the KEAP1-NRF2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service