Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

919721

Sigma-Aldrich

Methyl-13C, d1 alcohol

≥98 atom % D, ≥99 atom % 13C, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CDH2OH
CAS Number:
Molecular Weight:
34.04
UNSPSC Code:
12352101

isotopic purity

≥99 atom % 13C
≥98 atom % D

Assay

≥99% (CP)

form

liquid

mass shift

M+2

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service