Skip to Content
MilliporeSigma
All Photos(1)

Documents

699357

Sigma-Aldrich

4-(Chloroacetyl)morpholine

97%

Synonym(s):

2-Chloro-1-(4-morpholinyl)ethanone, 2-Chloro-1-morpholinoethanone, 2-Morpholino-2-oxoethyl chloride, 4-Morpholinylcarbonylmethyl chloride, N-(Chloroacetyl)morpholine, Chloroacetic acid morpholide, Chloroacetic acid morpholine amide, Morpholine chloroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10ClNO2
CAS Number:
Molecular Weight:
163.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.512

mp

26-30 °C

density

1.257 g/mL at 25 °C

SMILES string

ClCC(=O)N1CCOCC1

InChI

1S/C6H10ClNO2/c7-5-6(9)8-1-3-10-4-2-8/h1-5H2

InChI key

YMQRPXBBBOXHNZ-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service