923494
iPrMPhos
Synonym(s):
1-Diadamantylphosphino-1′-diisopropylphosphinoferrocene
About This Item
Recommended Products
form
powder or chunks
Quality Level
reaction suitability
reagent type: catalyst
reaction type: Cross Couplings
reagent type: ligand
mp
172-175 °C
functional group
phosphine
Related Categories
Application
Learn more about MPhos ligands and catalysts
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
YPhos ligands facilitate palladium-catalyzed coupling reactions of aryl chlorides under mild conditions with short reaction times.
YPhos ligands facilitate palladium-catalyzed coupling reactions of aryl chlorides under mild conditions with short reaction times.
YPhos ligands facilitate palladium-catalyzed coupling reactions of aryl chlorides under mild conditions with short reaction times.
YPhos ligands facilitate palladium-catalyzed coupling reactions of aryl chlorides under mild conditions with short reaction times.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service